PIG® Universal Absorbents
Absorbs anything, works everywhere and is the #1 brand worldwide.
STRONGEST MAT ON THE MARKET
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market. It won’t rip, tear or fray, even when saturated.
SOAKS IT ALL IN
Highly absorbent, fine-fiber construction absorbs oil, coolants, solvents and water without letting liquid or fiber residue behind. Absorbs quickly Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup.
FLAME-RESISTANT
Mat won’t burn immediately like untreated cellulose mats and melts when exposed to high heat .
KEEPS YOU COMPLIANT
Mat helps you comply with 29 CFR 1910.22(a)(2) for maintaining floors in a clean, dry condition.
Lightweight, 15" x 20", 200 pads. PIG Mat lasts 2x longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
Mediumweight, 15" x 20", 125 pads. PIG Mat lasts 2x longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
Lightweight, 15" x 20", 200 pads. PIG Mat lasts 2x longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
Mediumweight, 30" x 200', 1 roll. PIG Mat lasts 2x longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
Lightweight, 15" x 150', 1 roll. PIG Mat lasts 2x longer than ordinary mats for fewer change-outs. Absorbs everything. Works everywhere.
5" x 10', Absorbs up to 8 gal. per sock, 4 socks. Our heaviest absorbent sock is filled with vermiculite to prevent spills from spreading.
Universal, Absorbs up to 2 gal./pillow, 16 pillows. When the situation calls for more, our high-capacity pillows deliver the absorbency you need.
19"x15", 100 pds. Value priced MRO Hazmat roll - When the job doesn't require the quality of PIG Mat, use MRO to get the job done!
30"x150', 1 roll. Value priced MRO Hazmat roll - When the job doesn't require the quality of PIG Mat, use MRO to get the job done!