PerkinElmer NexION ICP-MS Series
For analytical testing laboratories conducting trace-elemental analyses, the NexION® ICP-MS Series from PerkinElmer builds on four decades of innovation in ICP-MS technology and a longstanding reputation for reliability, ease of use, and low maintenance—delivering exceptional performance, accuracy, and repeatability.
The NexION 1100 builds on a strong tradition of reliable, easy-to-use, low-maintenance instrumentation, it reinvents the user experience for routine ICP-MS applications.
The NexION 2200 builds on the 1100 with novel and proven technologies come together to deliver uncompromised performance, accuracy, and repeatability – and even more sensitivity.
The industry’s first four-quadrupole system taking ICP-MS performance beyond high-resolution ICP-MS and traditional triple-quad technology to deliver exceptionally low background equivalent concentrations (<1 ppt, even in hot plasma) and outstanding detection limits.