Industrial Microbiology
There is more to safety than meets the eye
Dehydrated Culture Media and Immunological Lateral Flow Tests for Traditional and Rapid Pathogen Detection.
Related Information:
Pathogen Testing: EMD Millipore Granulated Culture Media
Safer: Minimizes airborne toxins and allergens, allergic responses and workplace contamination
Accurate: Prevents component separation and clumping even under humid or warm conditions
Fast: Dissolves rapidly in water
Easier: Superior flow and non-sticking properties for easier handling
Reliable: Homogenous distribution of ingredients assures high reproducibility even with small media quantities
Pathogen Testing: Lateral Flow Tests
Reliable: Same accuracy standards as classical detection methods
Comprehensive: Tests cover six most relevant pathogens in food
Fast: Definite results within 20 - 30 minutes
Easy-to-use: Clear yes/no results after simple sample application
Safe: Additional positive control and specially adapted enrichment media for precise, reliable results
Economic: Rapid results save operating costs by expediting product release while ensuring product reliability
Microbial Air Monitoring Solutions
With active microbial air monitoring products from MilliporeSigma, you can be sure to get the most accurate sample solutions available. Our easy-to-use monitoring systems fully comply with all international standards and regulations – ensuring reproducible and reliable results.