Specialty Wipes and Cleaning Tools for Efficient and Ergonomic Sanitization
Process cabinents, curved surfaces, and spaces between, under, and behind equipment need special cleaning attention. Micronova's range of specialty cleaning tools and wipes simplify the task. From biosafety cabinets to lyophilizers, round vessels to curved joints, and plastic strip curtains to soft walls, there are options for removing contaminants for the most challenging of conditions.
MegaSwat™ Mops Blade Style Mop
Single ply and foam lined blade style mops easily glide under and behind equipment. Adapters and mop covers are available in a range of widths, lengths and materials.
Cleans both sides of a strip curtain at the same time allowing for a more efficient and effective cleaning.
Ergonomic and effective cleaning for isolators, hoods, RABS and other small cabinets. 360° swivel and available in a range of fabrics for cleaning or disinfection.
Cleans top and bottom of shelves simultaneously. Adjustable spring loaded plates provide consistent pressure and release of cleaning agent.
Use on glass or stainless walls or specialized floor mats. Dual Surface TMop
Use on glass or stainless walls or specialized floor mats. Dual Surface TMop™ adapter and squeegee. Apply solution with TMop adapter and dry finish with squeegee.
Flexible adapter conforms to tank with pressure against surface for good cleaning contact. Easily cleans tank and cylinder sides, bottoms, and both interiors and exteriors of vessels.