NovaClean™ Family of Cleanroom Detergents
Nonionic detergents remove particles and residues common in cleanrooms and critical laboratories. With low salts and metals, in the ppb range, these cleanroom detergents are ideal for semiconductor, electronics, medical device, and pharmaceutical industries.
An all-purpose concentrated cleanroom detergent with a fine filtration and purity that is designed for high tech industries, well suited for med device and biotech industries. Free rinsinig and biodegradable.
NovaClean™ AFX Amine-Free Floor Cleaner
NovaClean™ AFX has the same fine filtration as original NovaClean™ concentrated cleanroom detergent with the added feature of an amine-free formulation. Particularly well-suited for semiconductor applications. Free-rinsing and biodegradable.
NovaClean™ Lab & Glass Cleanroom Detergent
A non-solvent based detergent formulated to be applied directly to cleanroom work surfaces including glass, stainless steel, epoxy, acrylic, and painted surfaces. Free-rinsing and biodegradable.
NovaHol™ with the same pure formula as original NovaClean™ detergent, has added isopropyl alcohol for a faster dry time. NSF rated, category A1 (general cleaners).
MegaClean™ Heavy-Duty Cleaner
A non-flammable, heavy-duty cleaner and degreaser that offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Water soluble and biodegradable.