Room Bio-decontamination with Bioquell Technology
Ecolab utilizes Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number: 72372-1-86703) to help eliminate microbial contamination from surfaces. This sterilant is emitted in a vapor form through the use of Bioquell hydrogen peroxide vapor systems.
There are multiple solutions available for room bio-decontamination from Ecolab including mobile, integrated and fixed systems. For large spaces, multiple systems can be used as the process is scalable.
- Hydrogen peroxide vapor generators in fixed, integrated, or mobile formats
- Create validated cycles for 6-log sporicidal kill on exposed surfaces utilizing Bioquell Hydrogen Peroxide Sterilant (EPA Registration Number 72372-1-86703)
- Various room volume capacities for bio-decontamination. Multiple systems can be used to increase maximize volume achievable.
- Designed with RFID label technology for traceability of peroxide and auditing
- 21 CFR Part 11 audit trail compliance package available
- Integration testing for BMS and more complex systems
- Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
- Supports GMP compliance
A mobile, scalable system, the Bioquell ProteQ features wireless communication, built-in aeration, optional 21 CFR Part 11 audit trail and more.
Wall-mounted, automated, and compact, the Bioquell SeQure kills microorganisms on exposed surfaces in smaller rooms and spaces with validated cycles.
The Bioquell L-4 can bio-decontaminate enclosures, equipment, and more or, with additional accessories, be used for room bio-decontamination.