Isolator with integrated Bioquell bio-decontamination technology
The Bioquell Qube delivers efficiency, reliability, and ease of implementation to your aseptic environment. An aseptic environment is created within the Bioquell Qube using Ecolab’s integrated bio-decontamination technology with Bioquell Hydrogen Peroxide Sterilant (EPA registration number 72372-1-86703). While it creates a grade A / ISO 5 equivalent workspace in the isolator, the system can reside in a lower grade cleanroom saving gowning time and operating costs.
Unlike traditional stainless steel Isolators, the Bioquell Qube system is modular and made from a hardwearing plastic mold, offering distinct tangible benefits. Because of its manufactured method, production of the system is quicker, consistently crafted, ergonomically designed, and minimizes the risks of leaks associated with welds, joints, and gaskets found in other Isolators.
It offers seamless integration options for various applications. Ecolab experts work with clients to ensure their process can be implemented within the system. With the capability to work in up to two modules while incoming supplies are bio-decontaminated, the Bioquell Qube allows for a dynamic and highly efficient workflow.
The Bioquell Qube provides an ideal alternative to traditional stainless steel isolators as an efficient, reliable, and easy to implement system for the bio-pharmaceutical, laboratory research, and biotechnology industries. Some of the features and benefits of the Bioquell Qube are:
- Integrated with Ecolab Bioquell hydrogen peroxide vapor technology
- Validated cycles create 6-log sporicidal kill on exposed surfaces
- Utilizes EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution
- Creates validated Grade A / ISO 5 environment
- Customize to meet your working needs
- Configurations up to 6 gloves, 2 material pass-throughs or rapid transfer ports
- One inch tri-clover port(s) in any chamber
- Rapid transfer port options for aseptic transfer and more
- Integrated options such as sterility test pump from Merck-Millipore
- 4 levels of environmental monitoring to select from
- Does not require connection to HVAC
- Operate a Grade A / ISO 5 environment while hosting in Grade C/D location
- Operates at positive or negative pressure
- Designed with RFID label technology for traceability and auditing
- Integrated screen and camera for recording and displaying worksheets
- 21 CFR Part 11 audit trail compliance package
- Can be manufactured, installed and validated in 16 weeks from order date
- Bio-decontaminate incoming materials while working in another chamber
- No construction or electrical work required for installation
- Cycle recording (thermal printer and solutions for real time cycle data, including electronic records in pdf format)
- Supports GMP compliance
Create an aseptic workspace with Ecolab’s Bioquell Qube, a configurable isolator integrated with Bioquell hydrogen peroxide vapor technology.