Ecolab's Complete Cleaning and Disinfection Product Range
Ecolab offers a wide range of products to assist in your routine and daily cleaning regiments. Whether for CIP or within the walls of your cleanroom. From floor to ceiling, wall to wall, Ecolab has an offering to help ensure your contamination risk is minimized.
Ecolab COSA™
Ecolab COSA™ Alkaline Cleaners for Pharmaceutical Manufacturers are specifically designed to remove typical residues found in CIP applications such as oils, fats, proteins and other organic materials. For pigments and inorganic soils, additives can be incorporated to aid in removal. For greases, oils, and other hydrophobic materials, surfactants are needed to help break up and emulsify these difficult-to-clean substances. All Ecolab COSA™ alkaline cleaners are supported by a complete validation package, including techniques for determining acceptable residual levels and analytical methods for residue quantification in CIP applications. With a full range of alkaline cleaners at varying levels of alkalinity and surfactants, Ecolab can find the right product to eliminate your toughest soils without damaging your equipment, putting your employees at risk, or creating problems in your wastewater treatment facility.
Ecolab Klercide™
The Klercide™ range of routine and rotational biocides and disinfectants are an ideal choice for helping control microbial contamination most likely to be found in a cleanroom environment. Our range provides the required efficacy for a cleanroom environment and compatibility with cleanroom surfaces as well as rotation cleaning and residue removal products. Ecolab also offers a mophead, bucket system, a range of sterile dry wipes (known as Klerwipe), isolator cleaning tool and more.
The SteriShield™ Delivery System
An effective cleaning and disinfection program in controlled areas is critical to maintain product quality and safety in the pharmaceutical manufacturing industry. The SteriShield™ Delivery System’s (SDS) closed and validated trigger spray system ensures content sterility throughout use, making it an ideal solution for the cleaning and disinfection of critical areas like cleanrooms.
The Klercide™ range of routine and rotational biocides and disinfectants help control microbial contamination to protect your processes.
COSA™ Alkaline Cleaners are specifically designed to remove residues found in CIP applications such as oils, fats, proteins & other organic materials.