Non-Sterile Cleanroom Manufacturing
DuPont non-sterile cleanroom garments are available as individually packaged as well as bulk packaged depending on your needs. Tyvek®, microporous film and SMS deliver the particle barrier, durability and comfort in use.
Tyvek® IsoClean®
- Comfortable, lightweight and durable
- Individually packaged garments offered as sterile or clean-processed & sterile
- Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
- Serged or bound seam styles are available
ProClean®
- Made from a microporous composite fabric
- Offers protection against non-hazardous particulates and light liquid splash
- Lightweight and low linting
ProShield® 10
- SMS garments are optimized for comfort, softness and breathability
- Available in a variety of coverall styles in a choice of white, blue and gray
- Designed for a range of cleanroom manufacturing and controlled environment applications.
- Nonhazardous dry particle and light liquid splash applications