Chemical Resistance/Flame Resistance
- Cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments
- Intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols
- Designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event
- Will not ignite and continue to burn after the flame source is removed
- Excellent choice as a cost efficient cover to protect reusable FR garments
- Applications include: Steel Mills, Coal Mines, Maintenance Work, Refineries, Laboratories, Utilities
- Provides chemical and secondary flame protection in a lightweight garment
- Intended to be worn over primary flame resistant garments
- In the event that a flash fire occurs, they won't ignite and won't contribute to additional burn injury if appropriate flame-resistant personal protective equipment, such as DuPont™ Nomex®, is worn beneath
- Provides an effective barrier against a range of inorganic acids and bases as well as industrial cleaning chemicals and particles
- Ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire resistance clothing worn beneath them
- Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations
- Single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc
- Combines the trusted chemical protection of Tychem® and thermal/arc protection of Nomex® into a single layer garment
- Designed to help provide industrial workers and Hazmat responders with permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents and to provide escape time from flash fires and protection from electric arc
- Provides at least eight hours breakthrough protection against 17 of the 21 standard ASTM F1001 chemicals and at least 30 minutes of protection for more than 180 chemical challenges including chemical warfare agents
- Typical Applications: HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
- Composed of a patented, heavy-duty aluminized fabric made of a DuPont™ Nomex®/DuPont™ Kevlar® blend laminated to a multi-layer barrier film
- Unique combination of layers provides excellent physical strength and cut and abrasion protection
- Ideal for industrial chemical, HazMat response and military applications including chemical warfare agents