Orders LinkContactUsLinkComponent
AnalytikJenaUVPChemGel-955x690.jpg
Analytik Jena UVP Chem/Gel Studio Imagers
Analytik Jena offers a wide range of high-performance BioImaging Systems designed to fit researchers' specifications and budgets. All systems come in two different sizes and are equipped for chemiluminescence and fluorescence applications with a UV transilluminator, RGB LEDs, and white light. Systems are upgradeable for NIR applications with NIR lasers.
High-performance Gel and Blot Imagers | Avantor
Product selection
23747816-1-400x495.jpg
UVP ChemStudio touch
High-performance gel and blot imager.
23747815-1-400x495.jpg
Imager perfect for larger gels and blots with a 25 x 26 cm imaging area.
33557615-400x495.jpg
UVP GelStudio touch
Daily use high-performance gel documentation imager.
25467148-400x495.jpg
High-performance imager for large gels.