HSW® Soft-Ject® syringes
Henke-Sass Wolf’s (HSW) Soft-Ject syringes combine a lubricated synthetic-rubber piston with a tight, high-transparency barrel for fine dosing and a smooth draw and flow, plus a double seal for increased safety. Premium and disposable, these 3-part disposable syringes are made from non-toxic material and contain no latex, pyrogen, or PVC
- Available 1mL to 50mL in Luer slip, Luer lock, and catheter versions
- 10mL syringes with 0.2mL increments
- 50mL and 100mL syringes with an extended graduation for wider range of application
- Soft-Ject Luer-lock syringes are compatible with most commercially available syringe pumps