Biochemical assays involve purified proteins or other targets tested in solution with candidate compounds. They are a staple of early drug discovery and typically rely on straightforward mix-and-read protocols.
- Assay Plates (96, 384, 1536 wells): Multi-well plates are the backbone of HTS. Higher-density formats (384-well and especially 1536-well plates) allow more tests per run, boosting throughput and reducing reagent use. Many biochemical assays can be miniaturized into 384- or 1536-well volumes relatively easily. These plates must meet ANSI/SBS standards for automation and may be made of polystyrene or cyclic olefin with clear, white, or black walls depending on detection mode.
- Detection Reagents and Kits: Biochemical screens often use colorimetric, fluorescent, or luminescent readouts. Common examples include fluorescent substrates for enzyme activity, FRET/TR-FRET reagents, or luciferase-based HTS assay kits for measuring ATP or second messengers. In fact, fluorescence-, luminescence-, and absorbance-based assays remain among the most widely used HTS technologies. Selecting high-quality, sensitive detection reagents ensures signal can be detected even in tiny reaction volumes.
- Buffer and Compound Handling Consumables: These assays require reliable source plates for compound libraries (e.g. deep-well plates), reservoir plates for bulk reagents, and automation-friendly pipette tips for liquid handling. Low-retention, sterile robotic tips help accurately dispense microliter volumes and prevent cross-contamination. Proper plate seals or lids are also needed to prevent evaporation when working with miniaturized volumes.