Essential Consumables for Spatial Biology:
What Every Lab Needs for Spatial Transcriptomics & Proteomics
Spatial biology – encompassing spatial transcriptomics and spatial proteomics – is revolutionizing how scientists study tissues. By preserving the spatial context of gene and protein expression, these techniques enable insights that traditional bulk genomics or single-cell methods cannot achieve. Researchers can map where in a tissue specific genes are active or particular proteins are expressed, linking molecular data to histological architecture. This capability is more than just a technical marvel; it’s driving breakthroughs in research and drug discovery. In fact, as sequencing costs plummet, spatial biology is rapidly being adopted in pharmaceutical R&Dto reveal crucial spatial patterns of disease that inform new therapeutic targets. Major scientific journals have heralded this paradigm shift – Nature Methods named spatial transcriptomics the 2020 “Method of the Year” for its power to retain positional context in biology, and more recently highlighted spatial proteomics for exposing how proteins are organized in complex tissues.
Industry interest is surging as well – the global spatial biology market is projected to nearly double from $533 million in 2023 to almost $1 billion by 2029. Clearly, spatial omics has arrived as a must-have approach for advanced biology labs.
But what does it take to implement spatial transcriptomics or proteomics in your lab?
Successful spatial biology experiments require a specialized set of consumables at each stage, from the moment a tissue sample is collected to the final imaging or sequencing readout. Ensuring you have the right supplies on hand is critical for preserving sample integrity and obtaining high-quality data. From spatial transcriptomics kits and barcoded slides to reagents for spatial proteomics assays and imaging-compatible labware, every step comes with essential tools. Below, we provide a step-by-step breakdown of a typical spatial biology workflow – from tissue prep to sequencing/imaging – highlighting the essential consumables at each stage. Along the way, we’ll also discuss the value of sourcing these items through Avantor’s extensive catalog, which offers a wide selection of products, strong logistics support, and expertise for regulated lab environments. Whether you’re mapping gene expression across a tumor or profiling protein markers in tissue, consider this your checklist of spatial omics consumables that every lab needs to get started.
Step 1: Tissue Collection and Preparation – Preserving Spatial Context
All spatial biology workflows begin with obtaining a high-quality tissue sample and preserving its architecture. The goal of this step is to maintain both the tissue morphology and the integrity of biomolecules (RNA or protein) so that spatial information is not lost. Key consumables for tissue collection and stabilization include:
Fixation reagents
Chemical fixatives like 10% neutral buffered formalin or 4% paraformaldehyde (PFA) are essential for formalin-fixed, paraffin-embedded (FFPE) workflows Proper fixation prevents degradation and locks molecules in place. If working with fresh-frozen samples instead of FFPE, rapid freezing in isopentane or liquid nitrogen is used to preserve RNA – in that case, cryomolds and embedding media such as OCT compound are critical consumables. Regardless of method, using nuclease-free reagents and sterile supplies at this stage is recommended to protect RNA from degradation.
Tissue embedding supplies
For FFPE, labs will need tissue processing cassettes, paraffin wax, and mold accessories to embed the fixed tissue in blocks. For frozen samples, disposable base molds and OCT are used to create cryo-blocks. These consumables ensure the tissue can be sectioned later without losing spatial orientation.
Storage and transport containers
Sterile, RNase/DNase-free sample tubes or cryovials are important if tissues will be stored (e.g. in LN2 freezers) or shipped. Maintaining cold chain with insulated containers and dry ice for frozen tissues, or preservative solutions for fresh tissues, helps retain sample quality from collection to processing.
By investing in proper tissue prep consumables up front, labs set a solid foundation for spatial analyses. As a supplier, Avantor offers all the essentials for this stage – from high-grade fixatives to cryopreservation media and histology cassettes – ensuring that your tissue’s spatial information is preserved from the very start.
Step 2: Tissue Sectioning and Slide Preparation – Getting Samples on the Right Surface
Once tissues are preserved and embedded (either in paraffin or OCT), the next step is to cut thin sections and mount them onto slides. This sectioning stage requires precision and the right consumables to capture tissue morphology intact. Typically, a microtome or cryostat is used to slice the tissue into sections ~5–10 μm thick, which are then placed on slides for downstream staining or molecular processing.
Essential consumables for this step include:
Essential consumables for this step include:
Microtome blades and sectioning tools
Disposable steel blades (for microtomes) or carbide knives (for cryostats) are needed to repeatedly slice tissues. Ensuring sharp, high-quality blades is key to getting intact sections without tears or folds. Brushes, forceps, and spatulas are also used to handle delicate sections and position them on slides.
Adhesive or coated slides
Standard microscope slides often require a coating (e.g. poly-L-lysine or charged slides) to help tissue sections adhere, especially for FFPE sections that will undergo washes. Having slides designed for tissue adhesion prevents sections from detaching during processing. Many labs use “plus” slides or silane-coated slides for this purpose – these are common lab consumables available through Avantor in bulk packs.
A unique consumable for spatial transcriptomics workflows is the barcoded slide. Unlike plain glass slides, these specialized slide kits have an array of spatially encoded oligonucleotides printed on their surface. When a tissue section is placed on such a slide, any mRNA captured from that tissue inherits a positional barcode from the slide. This technology, first introduced by Ståhl et al. (2016) and commercialized in platforms like 10x Genomics Visium, enables mapping of gene expression to coordinates on the tissue. In practice, these slides come with pre-spotted capture probes (often poly(dT) to bind mRNA) in a grid pattern. Each spot on the slide has a unique barcode, so transcripts from cells in one location are tagged differently than those just 100 microns away. If your lab is performing spatial transcriptomics, obtaining the appropriate barcoded slide kit is mandatory. Avantor can help source these specialized consumables (often sold as part of spatial transcriptomics kits) and ensure you have the correct format for your platform – an important consideration, since slides are usually vendor-specific. For labs using emerging DIY methods (like printing your own oligo arrays), Avantor also offers microarray slides and reagents to create custom barcoded surfaces.
Slide labels and storage
Maintaining sample identity and integrity is crucial. Labs should have durable barcode labels for slides (especially if processing many samples) and slide storage boxes or humidity-controlled chambers to hold sections before use. Keeping sections at proper temperature (e.g. in a 4°C fridge for short-term, or -80°C if long-term storage of slides with tissue) often requires sealed slide mailers and desiccants to avoid moisture.
By the end of this step, you should have tissue sections securely mounted on suitable slides – whether standard coated glass or advanced barcoded arrays. The quality of sectioning and mounting will directly impact downstream staining and image analysis, so using reliable consumables here pays off. Avantor’s portfolio includes a wide selection of microscope slides and sectioning supplies, meaning labs can easily obtain everything needed for this foundational stage through a single provider.
Step 3: Sample Pretreatment – Preparing Sections for Molecular Analysis
With tissue sections on slides, a series of pretreatment steps is usually necessary before you can interrogate RNA or proteins in the tissue. The goal of pretreatment is to make targets accessible while retaining tissue morphology. The exact steps depend on the sample type:
For FFPE sections
Slides must be deparaffinized and rehydrated. This requires consumables like xylene, ethanol series (100%, 95%, etc.), and distilled water or buffers. After removing paraffin, an antigen retrieval buffer is typically applied (e.g. citrate buffer pH 6 or Tris-EDTA pH 9) and heated to undo crosslinks formed by formalin fixation, thus exposing protein epitopes or nucleic acids. These retrieval solutions (available as ready-to-use buffers or concentrates) are critical for successful staining downstream. In addition, FFPE samples often need a few washes in phosphate-buffered saline (PBS) or similar buffers to equilibrate the tissue. Avantor supplies high-purity solvents and buffer reagents suitable for histology and retrieval steps, ensuring consistency in this critical process.
For fresh frozen sections
There is no paraffin to remove, but sections are typically fixed on-slide to stabilize them. For example, a common protocol is to post-fix cryosections in cold acetone or 100% methanol, or a brief PFA fix, to keep tissue morphology. After fixing, sections might be rinsed in PBS. Additionally, permeabilization reagents (like 0.1–0.5% Triton X-100 detergent, or enzymatic digestion in some protocols) may be applied to allow antibodies or probes to penetrate cells. If the spatial workflow involves in situ hybridization, a proteinase or other digest enzyme is sometimes used to expose target RNA within the tissue.
Nuclease-free conditions
Whether FFPE or fresh, when RNA will be analyzed, it’s vital to use RNase-free water, buffers, and consumables at this stage. For instance, using molecular biology grade water, filtered tips, and dedicating solutions to avoid RNase contamination will protect the integrity of transcripts. As noted in spatial protocol guides, even slide-handling tools should be clean and preferably RNase-free. Many labs include an RNase decontamination step for equipment and wear gloves to further safeguard RNA. Avantor’s catalog includes nuclease-free tubes, reagents, and certified clean labware to support these best practices.
During pretreatment, having the right reagents on hand ensures your tissue section is optimally prepared for the sensitive detection steps to follow. Skipping proper deparaffinization or using subpar buffers can lead to poor staining or loss of RNA signals. By sourcing high-quality solvents and molecular-grade reagents through a trusted supplier like Avantor, labs can perform these prep steps confidently and reproducibly. Remember, preserving both the tissue histology and molecular integrity is key at this stage, and the right consumables make all the difference.
Step 4: Target Labeling – Hybridization and Staining Reagents for Spatial Analysis
With tissues mounted and pretreated, the core analytical step of spatial biology involves labeling the targets of interest – whether those are RNA transcripts (for spatial transcriptomics) or proteins (for spatial proteomics). This step typically uses specialized kits and reagents to either hybridize to nucleic acid targets or stain proteins in situ. It’s arguably the heart of the workflow, and thus having the appropriate consumables is paramount. We’ll break this into two modes of target labeling:
4A. Labeling RNA Targets (Spatial Transcriptomics):
For spatial transcriptomics experiments, especially those aiming to profile many genes at once, the strategy often involves capture probes or hybridization of oligonucleotides in situ:
Sequencing-based spatial transcriptomics
If you are using a sequencing-based platform (like the barcoded slides mentioned earlier), the tissue on the slide will be subjected to a series of enzymatic reagents that capture and tag RNA. Permeabilization buffers are applied to the tissue to lyse cells just enough to release mRNAs onto the slide. Then a reverse transcription kit is used on-slide: it contains enzymes (reverse transcriptase), primers (often the barcoded oligo on the slide acts as a primer), dNTPs, etc. This process creates cDNA from the captured mRNA, incorporating spatial barcodes and unique molecular identifiers (UMIs). After first-strand synthesis, additional reagents are used to denature and collect the cDNA, which then goes into a library prep kit. All these reagents – permeabilization enzymes, RT mix, amplification and library prep kits – are specialized consumables. They often come bundled in spatial transcriptomics kits provided by platform manufacturers. Avantor helps labs by offering a variety of these kits and related molecular biology reagents, so you can obtain your reverse transcriptase, amplification enzymes, and indexing primers through a single source. Even if you’re assembling a custom workflow, you’ll need high-quality PCR enzymes, purification beads, and indexing adapters as consumables for this labeling process.
Probe-based hybridization (targeted spatial transcriptomics)
An alternative approach is using hybridization panels to detect a predefined set of genes in situ. Techniques like RNAscope (RNA in situ hybridization) or multiplexed FISH methods fall in this category. In these workflows, you will use a kit of target-specific probes that hybridize directly to mRNA within the tissue section. For example, the MERFISH technique uses hundreds of fluorescent probes to bind transcripts, and then reads them out via microscopy. Vizgen’s MERSCOPE platform provides gene panels and hybridization buffers as consumables, enabling detection of up to 500 genes with subcellular resolution . Essential consumables here include the probe set (often a mixture of DNA oligonucleotides designed for your genes of interest), hybridization buffer, wash buffers, and sometimes amplification reagents or fluorescent readout probes. These kits may also come with positive control probes and mounting media optimized for the assay. Avantor can assist in sourcing in situ hybridization kits or custom probe libraries, connecting researchers with leading products for spatial omics assays. By using a validated hybridization kit, labs save time on probe design and can trust that the reagents will produce specific, multiplexed detection of RNA targets in the tissue.
4B. Labeling Protein Targets (Spatial Proteomics):
For spatial proteomic analysis, the focus shifts to detecting proteins in situ, typically through immunohistochemistry (IHC) or immunofluorescence (IF) techniques. The consumables needed are those common to advanced histology and some unique reagents for multiplexing:
Primary antibodies
High-quality primary antibodies against the proteins of interest are the cornerstone of spatial proteomics experiments. Depending on the study, you might label a handful of markers or deploy large panels of 10–40 antibodies for multiplex imaging. It’s important to source antibodies that are validated for IHC/IF on tissue sections (often denoted by the manufacturer). Avantor’s catalog includes a vast array of antibodies from various suppliers, so researchers can find the clones specific to their targets. For multiplex assays, consider using primary antibodies from different host species or different isotypes, which enables simultaneous detection.
Detection reagents (secondary antibodies or conjugates)
Once primaries are bound, you need a way to visualize them. Secondary antibodies conjugated to fluorophores are commonly used for IF. For example, anti-mouse IgG tagged with Alexa Fluor® dyes, or anti-rabbit IgG with FITC, etc., depending on the primary. If doing chromogenic IHC, secondary antibodies conjugated to enzymes like HRP are used along with a DAB substrate kit (diaminobenzidine chromogen) to produce a colored precipitate. In multiplex fluorescence protocols (like cyclic immunofluorescence), you might use directly labeled primary antibodies or DNA-barcoded antibodies to allow iterative or simultaneous detection. All these are consumables to plan for: fluorescent antibody kits, enzyme substrate solutions, mounting media with antifade, and even spectral calibration slides for some imaging systems. Avantor provides a broad selection of immunochemistry reagents – from single-color secondary antibodies to complete multiplex IHC kits – so labs can get all necessary detection reagents with assured compatibility.
Buffers and blocking reagents
Immunostaining workflows require several buffers (often Tris or PBS-based) for washing off unbound antibodies between steps. Additionally, blocking buffers (e.g. normal serum, BSA, or specialized protein blocking solutions) are essential consumables to prevent nonspecific binding. These reagents improve signal-to-noise by reducing background. It’s important to have buffer components that are high purity and tested for immunolabeling. Many suppliers (accessible via Avantor) offer ready-made IHC buffer packs and blocking solutions to streamline this step.
Histological stains and counterstains
To complement molecular staining, labs often use traditional stains to provide tissue context. Hematoxylin and eosin (H&E) is commonly used either in a serial section or even on the same section (some workflows perform an H&E after transcriptomic probe extraction). A nuclear counterstain like DAPI (4’,6-diamidino-2-phenylindole) is frequently applied in fluorescent protocols to mark nuclei. These dyes are standard consumables: H&E kits, DAPI or Hoechst stain, and other specialized dyes (e.g. a collagen stain for extracellular matrix) can be included to enrich the spatial data. Avantor carries a variety of biological stains and dye reagents suitable for such purposes.
In summary, the target labeling step in spatial biology makes use of sophisticated kits and reagents – from gene probe sets and amplification mixes to antibody cocktails and staining solutions. Each laboratory will choose the approach that fits their research question (transcripts vs proteins or even both), but in all cases, sourcing high-quality, validated reagents is vital. Using suboptimal or expired reagents here could mean weak signals or false readings that undermine the entire experiment. By procuring these critical consumables through Avantor, labs gain access to reliable products from top brands and can ensure batch-to-batch consistency (important for multi-slide studies). The ability to get spatial transcriptomics kits, multiplex immunofluorescence reagents, and other spatial omics consumables from one supplier also simplifies inventory management and protocol planning.
Step 5: Detection and Data Acquisition – Imaging Systems, Sequencing, and Compatible Labware
After labeling the targets in the tissue, the final step is to collect data that captures both the molecular information and its spatial coordinates. Depending on the spatial workflow, this data acquisition is done either by imaging the tissue (for methods that use visual readouts) or by sequencing the extracted molecules (for NGS-based methods). While the microscopes or sequencers themselves are instruments rather than consumables, there are several consumable items and labware needed to successfully go from labeled tissue to analyzable data:
Microscope slides, cover slips, and imaging labware
If your readout is imaging (fluorescence microscopy, for instance), having the right imaging-compatible labware is essential. This includes high-quality cover slips (often #1.5 thickness coverslips are used for optimal optical clarity under high-magnification lenses) and mounting media. An anti-fade mounting medium is a critical consumable when imaging fluorescently labeled tissues – it preserves signal and prevents photobleaching of fluorophores during imaging. Additionally, some labs use specialized chambered imaging slides or multi-well plates with optical glass bottoms for automated high-content imaging systems. These allow multiple samples or conditions to be imaged in one run. Avantor provides a range of microscope slides and cover glasses that are guaranteed to be free of distortions and autofluorescence, which is important for clear images. If using a digital slide scanner for whole-slide imaging, make sure to use slides of the recommended dimensions and thickness for the instrument, and consider calibration slides (with known fluorescence or color patterns) as consumables to verify the imaging system’s performance.
Sequencing library prep kits and reagents
For sequencing-based spatial transcriptomics, once cDNA is collected from the slide (as described in Step 4A), it needs to be converted into a sequencing library. This involves end repair, adapter ligation, PCR amplification, and adding indices – steps very similar to standard RNA-seq library preparation, but with the twist that the cDNA already carries spatial barcodes. Kits for NGS library preparation are therefore essential consumables. Depending on the platform, you might use a specific kit provided by the spatial technology vendor, or a generic DNA library prep kit (compatible with Illumina sequencers, for example). Cleanup beads (AMPure or equivalents), quantification reagents (like Qubit DNA assay kits or Bioanalyzer chips), and quality control supplies are also needed at this stage. Avantor’s genomics product selection includes many of these kits and reagents, ensuring you can reliably build libraries ready for sequencing.
Sequencing consumables
Finally, to sequence the libraries and get digital gene expression data with spatial barcodes, you will use consumables tied to the sequencer. For an Illumina sequencer, this means a sequencing reagent kit (flow cell and reagents for clustering and sequencing by synthesis) and perhaps custom sequencing primers if required by the assay (some spatial protocols use custom read primers to read the barcode and UMI sequences). These kits are typically obtained from the sequencer manufacturer, but they are an important part of your consumable planning and can often be sourced through distributors like Avantor for convenience. Ensure you have the correct read length configuration – for example, spatial transcriptomics libraries often require paired-end reads (one read for the gene insert, another for the spatial barcode). Sufficient capacity should be allocated (number of reads per spot or per ROI) so having extra sequencing flow cells on hand is wise if repeats are needed.
Data storage and analysis tools
While not a traditional consumable, it’s worth noting that the massive images or sequencing data files generated will require robust data storage solutions and analysis software. Make sure to budget for high-capacity storage or cloud services, and consider bioinformatics support. Avantor, in addition to consumables, also offers informatics and software solutions in some cases, or can connect labs with the right services to handle spatial data. Although data analysis is beyond the scope of lab consumables, being prepared with these resources ensures the effort spent on reagents and experiments translates into actionable results.
At the end of this detection step, you will have either rich multi-channel images of your tissue or a sequencing dataset representing spatially resolved gene expression. The labware and consumables you used in this phase contribute to the fidelity of your data – blurry images can result from using cheap coverslips or improper mounting medium, and sequencing quality can suffer if library prep reagents are not up to par. By choosing high-quality imaging consumables and trusted NGS kits via Avantor, labs improve their chances of obtaining publication-ready spatial data on the first go. Plus, Avantor’s logistics capabilities (including cold-chain shipping for kits and just-in-time delivery options) mean that even these sensitive reagents arrive when and where you need them, in optimal condition.
The Avantor Advantage – Streamlining Spatial Workflows with One-Stop Sourcing
Each step of the spatial biology workflow requires a variety of reagents and materials, and procuring them from different vendors can be a hassle. This is where Avantor’s value truly shines. As a leading global supplier of laboratory products, Avantor (and its VWR® portfolio) offers comprehensive product categories covering everything mentioned in the workflow above – and more. By leveraging Avantor as your single sourcing partner, you can simplify procurement and ensure quality across the board. Here are some key advantages of choosing Avantor for your spatial biology consumables:
Wide Selection of Products
Avantor provides a broad, reliable supply of laboratory consumables that span genomics, proteomics, and histology needs. This means you can find everything from basic lab staples (slides, solvents, pipettes) to advanced spatial transcriptomics kits in one place. Need RNA-friendly microtome blades, a specific hybridization buffer, or a set of fluorescent secondary antibodies? Chances are Avantor has multiple options available. This wide selection not only offers convenience but also allows you to compare alternatives (different brands or grades) and choose what best fits your experimental requirements and budget.
Logistics Expertise and Reliable Delivery
Running cutting-edge spatial experiments often means coordinating complex timelines and maintaining reagent conditions (many kits are temperature-sensitive). Avantor’s logistics expertise ensures that your critical reagents arrive on time and in optimal condition. The company’s global distribution network and inventory management can be a lifesaver for labs on tight schedules – for instance, getting that last-minute order of fixative or overnight shipping of a replacement kit if something goes wrong. Avantor is experienced in cold-chain shipping for enzymes and antibodies, and can consolidate orders to reduce the number of shipments you have to track. This level of service lets researchers stay focused on the science instead of worrying about supply hiccups.
Quality and Compliance Support
For labs in regulated environments (clinical labs, pharma/biotech organizations under GLP/GMP), sourcing support is crucial. Avantor offers extensive sourcing support for regulated labs, meaning they can provide documentation like certificates of analysis, lot traceability, and even assist in finding compliant-grade reagents (e.g. molecular biology reagents that meet clinical diagnostic standards or GMP-manufactured chemicals when required). Having a supplier that understands regulatory compliance can ease the validation burden when introducing spatial assays in a clinical trial or a diagnostic setting. Additionally, Avantor’s stringent quality control for its own brands and the reputable manufacturers it partners with gives confidence that each batch of consumables will perform consistently – a key factor for reproducibility in spatial studies.
Technical Expertise and Customer Support
Avantor isn’t just a catalog; it’s also a resource. Their representatives and scientific support teams can help guide you to the right product for your application. Unsure which slides are best for an RNAscope assay, or which imaging plates work with your high-content imager? Avantor’s team can leverage their product knowledge to make recommendations. This kind of consultative support is especially valuable in a fast-evolving field like spatial biology, where new products and techniques are constantly emerging. Avantor keeps tabs on these developments (for example, stocking the latest spatial omics kits or updated formulations of reagents) so that customers have access to cutting-edge tools soon after they’re available.
In short, partnering with Avantor means streamlined workflows and peace of mind. You can equip your lab with all the essential spatial biology consumables knowing they’ll be of high quality and delivered efficiently. This one-stop shopping and support reduce downtime and uncertainty, allowing you to focus on experimental design and data analysis – the things that truly progress your research.
Conclusion and Call to Action: Equip Your Lab for Spatial Success with Avantor
Spatial transcriptomics and proteomics are powerful approaches opening new frontiers in biomedical research. From mapping tumor immune cell neighborhoods to discovering novel biomarkers, these techniques provide a level of insight that is driving innovation in drug discovery and beyond. However, as we’ve detailed, running spatial biology experiments requires careful coordination of many moving parts – and having the right consumables at every step. By understanding the workflow and securing essential items like barcoded slides, fixation and staining reagents, hybridization kits, and imaging labware, your lab will be well-prepared to generate high-resolution spatial data with confidence.
Avantor is committed to supporting researchers in this spatial revolution. With its extensive product portfolio and deep expertise, Avantor makes it easy to find and obtain all the spatial omics consumables you need to get started or scale up. We encourage you to take advantage of Avantor’s resources as you plan your spatial experiments. Explore Avantor’s spatial biology product offerings – visit the Avantor website or contact our specialists to discover how we can help outfit your lab for spatial transcriptomics and proteomics success. Whether you’re looking for a complete spatial transcriptomics kit, specific reagents for spatial proteomics, or simply advice on what consumables fit your workflow, Avantor is here to accelerate your journey into spatial biology. Equip your lab with the essentials and unlock new dimensions in your research!