My Account Menu
United States
My Account Menu
liquid-silicone-rubbers-hero_rdax_1170x548_80.jpg
Liquid silicone rubbers
NuSil® liquid silicone rubbers (LSRs) provide an excellent medical-grade material for precision-molded components, such as O-rings, gaskets, valves and seals used in medical devices.
Shop now
NuSil liquid silicone rubbers (LSRs) from Avantor® have a paste-like consistency and are designed for use in high-volume precision manufacturing processes such as liquid injection molding and over-molding. They can be used to produce molded parts that feature complex geometries in a matter of seconds, especially very small to medium-sized parts, like gaskets and valves used in medical devices.
LSRs are two-part platinum-catalyzed systems that cure rapidly with increased temperatures and do not require a post-cure process. Specialty formulations are available for applications that require solvent-resistance, high-clarity or lubrication. LSRs can be supplied in both high-volume containers for full-scale production or small handheld cartridges for prototyping or small volumes.
For additional functionality, LSRs can easily incorporate additives for color, radiopacity or to create a foam-like texture.
Custom formulations tailored to your needs
nusil-quality-375x235.jpg
NuSil has the expertise, processes, and proprietary technology to customize our silicones based on your unique specifications. We leverage our insight into the unique silicone chemistry and material properties to supply off-the-shelf and custom formulations tailored to your process and end-application performance requirements.
Learn more about how we can help you get the products you need.
Contact us
Product finder
medical-implants-2-375x235.jpg
NuSil’s liquid silicone rubbers are an excellent option for the high-throughput production of components used in long-term (> 29 days) implantable medical devices.
general-healthcare-375x235.jpg
When fabricating components such as gaskets and valves for short-term implant (< 29 days) or external medical devices, NuSil medical-grade LSRs provide an efficient, customizable material for high-throughput automated production.
Related content
Browse additional resources related to liquid silicone rubbers.
How can we help you?
Whether you’re using NuSil HCRs to extrude tubing or LVEs to encapsulate electronics, NuSil experts will help you determine the right silicone for your application.
CONTACT US