Description
- High purity; <0.05% D4 cyclic content
- Proprietary process creates an elastomer gel free of pilling and balling
- High clarity silicone
- Enhances soft focus attributes and increased cushion
- Mattifying properties
- Non-tacky film former that provides excellent coverage
- Texturizing balance – adds structure while not compromising spreadability
- Ideal for non-irritating and sensitive skin care applications
Applications
- Moisturization
- Anti-Aging/Anti-Wrinkle
- Sunscreen, After- Sun, and Self- Tanners
- Skin Repair Serums
- Skin Tone Repair
- Men’s Grooming
- Color Cosmetics
Product Details
Property | Average Result |
---|---|
INCI Name | Cyclopentasiloxane (and) Dimethicone/Vinyl Dimethicone Crosspolymer |
D4 Content | <0.05 % |
Viscosity | 550,000 cP (550,000 mPa*s) |
Appearance | Clear |
Non Volatile Content | 12 % |
Refractive Index | 1.40 |
Specific Gravity | 0.96 |
Comment | 12% Solids Elastomer Gel Dispersed in D5 |