Description
- Oil-in-water suspended gel
- Surfactant Free, Amodimethicone Free, Propylene Glycol Free, Chelating Agent Free, Silicone Membrane Free
- Silicone is stably suspended in the aqueous media without separation over time
- Ease of use: dispersed directly in the water phase
- Transformable texture: At first, a watery feel on one surface and a creamy texture on the other, both transforming into a matte cushiony feel
- Possible to formulate transparent cosmetic formulations
Applications
- Moisturization
- Anti-Aging/Anti-Wrinkle
- Sunscreen, After-Sun, and Self-Tanners
- Skin Repair Serums
- Skin Tone Repair
- Men’s Grooming
- Color Cosmetics
Product Details
Property | Average Result |
---|---|
INCI Name | Aqua (And) Dimethicone (And) Dimethicone/Vinyl Dimethicone Crosspolymer (And) Dimethiconol (And) Butylene Glycol (And) Carbomer (And) Phenoxyethanol (And) Sodium Hydroxide |
D4 Content | <0.05 % |
Viscosity | 165,000 cP (165,000 mPa*s) |
Appearance | White |
Refractive Index | 1.40 |
Specific Gravity | 0.98 |
Comment | Suspended Silicone with 30% CXG-1104 and 7% CSF-3504 |